Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide Reagent1L
Catalog No. 502901942
Encompass
Dimethyl sulfoxide is an amphipathic molecule soluble in both aqueous and organic solvents. It is commonly used as an aprotic solvent and also as a reagent in organic synthesis.
Catalog No. 50-290-1942 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D58791L
Dimethyl sulfoxide is an amphipathic molecule soluble in both aqueous and organic solvents. It is commonly used as an aprotic solvent and also as a reagent in organic synthesis.