Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide suitabl
Catalog No. 502898390
Encompass
Dimethyl sulfoxide (DMSO) is a polar organic solvent. DMSO activated by various electrophiles (oxalyl chloride) has been used to oxidize various alcohols (primary secondary allylic benzylic hindered and bicyclic). Carbonyl compounds are formed as reaction products. Due to its higher stability it has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction.
Catalog No. 50-289-8390 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348692L
Dimethyl sulfoxide (DMSO) is a polar organic solvent. DMSO activated by various electrophiles (oxalyl chloride) has been used to oxidize various alcohols (primary secondary allylic benzylic hindered and bicyclic). Carbonyl compounds are formed as reaction products. Due to its higher stability it has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction.