Learn More
Dimethyl Sulfoxide, Tested according to Ph.Eur., Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Solstice has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Density | 1.10g/mL |
| Quantity | 100 mL |
| Explosion Limit | 42% (63°F) |
| Flash Point | 87°C (188.6°F) |
| Ignition Point | 573°F |
| Packaging | Glass Bottle |
| Linear Formula | (CH3)2SO |
| Vapor Pressure | 0.42mmHg (20°C) |
| Beilstein | 506008 |
| Formula Weight | 78.13g/mol |
| Show More |
Safety and Handling
| RTECSNumber | 200-664-3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.