Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, USP, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18602242 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-242 500 mL Amber Glass Bottle
18-602-241 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-242 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1258500ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D1258, 67-68-5, C2H6OS

Spectrum™ Chemical Dimethyl Sulfoxide, USP is also known as DMSO. It is used topically to decrease pain and speed the healing of wounds, burns, and muscle and skeletal injuries. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

CAS Max % 100%
Boiling Point 189°C/372.2°F
Packaging Amber Glass Bottle
Refractive Index 1.4755-1.4775
Quantity 500 mL
Specific Gravity 1.095-1.101
Percent Purity 99.9%
Grade USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Dimethyl Sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.