Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Tetrachloroterephthalate 96.0+%, TCI America™
SDP

Catalog No. T106425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T106425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T106425G Supplier TCI America Supplier No. T106425G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1861-32-1
Molecular Formula C10H6Cl4O4
Molecular Weight (g/mol) 331.95
MDL Number MFCD00014902
InChI Key NPOJQCVWMSKXDN-UHFFFAOYSA-N
Synonym Tetrachloroterephthalic Acid Dimethyl Ester
PubChem CID 2943
ChEBI CHEBI:34664
IUPAC Name 1,4-dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl

Specifications

Physical Form Powder/Crystals
Melting Point 156°C
Quantity 25 g
Formula Weight 331.95
Percent Purity ≥96.0% (GC)
Chemical Name or Material Dimethyl Tetrachloroterephthalate
RTECSNumber WZ1500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.