Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Thiazolidine-2,4-dicarboxylate 98.0+%, TCI America™
SDP

Catalog No. T14525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14525G Supplier TCI America Supplier No. T14525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 318233-97-5
Molecular Formula C7H11NO4S
Molecular Weight (g/mol) 205.228
MDL Number MFCD00191687
InChI Key ZZAHPKXSIYXTAP-UHFFFAOYSA-N
Synonym Thiazolidine-2,4-dicarboxylic Acid Dimethyl Ester
PubChem CID 2763763
IUPAC Name dimethyl 1,3-thiazolidine-2,4-dicarboxylate
SMILES COC(=O)C1CSC(N1)C(=O)OC

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 5 g
Formula Weight 205.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Thiazolidine-2,4-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.