Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl trans-Stilbene-4,4'-dicarboxylate 98.0+%, TCI America™
SDP

Catalog No. S0502250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0502250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0502250MG Supplier TCI America Supplier No. S0502250MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34541-73-6
Molecular Formula C18H16O4
Molecular Weight (g/mol) 296.322
InChI Key JOODVYOWCWQPMV-ONEGZZNKSA-N
Synonym e-dimethyl 4,4'-ethene-1,2-diyl dibenzoate, dimethyl 4,4'-stilbenedicarboxylate, dimethyl trans-stilbene-4,4'-dicarboxylate, dimethyl stilbene-4,4'-dicarboxylate, benzoic acid, 4,4'-1e-1,2-ethenediylbis-, dimethyl ester, 4,4'-bis carbomethoxy stilbene, methyl 4-1e-2-4-methoxycarbonyl phenyl vinyl benzoate, methyl 4-e-2-4-methoxycarbonyl phenyl ethenyl benzoate, dimethyl 4,4/'-stilbenedicarboxylate
PubChem CID 5378468
IUPAC Name methyl 4-[(E)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC

Specifications

Melting Point 238°C
Color White-Yellow
Quantity 250 mg
Formula Weight 296.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl trans-Stilbene-4,4′-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.