Learn More
Medchemexpress LLC Dimethyl trisulfide | 3658-80-8 | 99.7% | 1 ML
SDP
Catalog No. 5000342011
Encompass_Preferred
Dimethyl trisulfide is a highly effective cyanide scavenger, naturally isolated from various plants including garlic, onions, and broccoli. This compound plays a crucial role in converting cyanide into thiocyanate. Beyond its chemical applications, it is recognized for its ecological significance, being utilized by saprophagous insects to pinpoint breeding grounds in decaying organic matter and by certain flowers to attract these insects.
  • Converts cyanide to thiocyanate, exhibiting over 40 times higher efficacy than thiosulfate.
  • Induces significant electrophysiological responses in insect antennae, aiding in ecological interactions.
  • Demonstrates protective effects against cyanide poisoning in in vivo studies.
  • Can be isolated from common plants like garlic, onions, and broccoli.
Catalog No. 50-003-42011 Supplier Medchemexpress LLC Supplier No. HY12845410MM/1ML
Dimethyl trisulfide is a highly effective cyanide scavenger, naturally isolated from various plants including garlic, onions, and broccoli. This compound plays a crucial role in converting cyanide into thiocyanate. Beyond its chemical applications, it is recognized for its ecological significance, being utilized by saprophagous insects to pinpoint breeding grounds in decaying organic matter and by certain flowers to attract these insects.
  • Converts cyanide to thiocyanate, exhibiting over 40 times higher efficacy than thiosulfate.
  • Induces significant electrophysiological responses in insect antennae, aiding in ecological interactions.
  • Demonstrates protective effects against cyanide poisoning in in vivo studies.
  • Can be isolated from common plants like garlic, onions, and broccoli.