Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl trisulfide, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019753 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9753 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9753 Supplier MilliporeSigma Supelco Supplier No. 7959210MGF
Add to Cart
Add to Cart

Dimethyl trisulfide is a potent odor active compound, which is found to be volatile in nature. It may be obtained via Strecker degradation of methionine under thermal conditions or from dimethyl disulfide.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 3658-80-8
Density 1.202 g/mL (at 25°C)
Flash Point 56°C - Closed Cup
Physical Form Neat
Linear Formula CH3 SSSCH3
MDL Number MFCD00039808
Melting Point -68°C
Boiling Point 58°C (15 mmHg)
Type Analytical Standard
Quantity 10 mg
Formula Weight 126.26
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.