Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Trisulfide 98.0+%, TCI America™
SDP

Catalog No. D341825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D341825G 25 g
D34185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D341825G Supplier TCI America Supplier No. D341825G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3658-80-8
Molecular Formula C2H6S3
Molecular Weight (g/mol) 126.25
MDL Number MFCD00039808
InChI Key YWHLKYXPLRWGSE-UHFFFAOYSA-N
Synonym dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
PubChem CID 19310
ChEBI CHEBI:4614
IUPAC Name (methyltrisulfanyl)methane
SMILES CSSSC

Specifications

Melting Point -68°C
Color Yellow
Boiling Point 170°C
Quantity 25 g
UN Number 1993
Formula Weight 126.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Trisulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.