Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Yellow Indicator, Solstice

Catalog No. 60046205 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-205 25 g Glass Bottle
60-046-204 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-205 Supplier Solstice Supplier No. 3312925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00008308
InChI Key JCYPECIVGRXBMO-UHFFFAOYSA-N
PubChem CID 6053
ChEBI CHEBI:17903
IUPAC Name N,N-dimethyl-4-phenyldiazenylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 111°C
Odor Odourless
Packaging Glass Bottle
Linear Formula C6H5N=NC6H4N(CH3)2
UN Number UN2811
Beilstein 746016
Color Yellow
pH 5 to 7 (at 10.00 g/L 20°C)
Solubility Information Insoluble in water
Quantity 25 g
Formula Weight 225.29g/mol
Grade Indicator
Chemical Name or Material Dimethyl Yellow
Show More Show Less
Hazard Category Danger
Hazard Statement P281-P301 + P310
Precautionary Statement H301-H351
DOTInformation Class 6.1 - PG 3 - RQ - Toxic solids, organic, n.o.s. HI: all
EINECSNumber 200-455-7
RTECSNumber BX7350000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.