Learn More
Medchemexpress LLC β,β-dimethylacrylshikonin (Isoarnebin I) | 24502-79-2 | MFCD00143687 | 99.3% | 370.40 | C21H22O6 | 10 MG
SDP
Catalog No. 5000289576
Encompass_Preferred
β,β-Dimethylacrylshikonin (Isoarnebin I) is a naphthoquinone derivative used in biomedical research for its anticancer and angiogenesis-modulating activities. It is supplied as a solid with high reported purity for laboratory experiments and is typically handled and stored protected from light.
  • High purity (99.3%) appropriate for research use.
  • Solid form with brown to reddish brown appearance.
  • Available in small research pack size (10 mg) for assay development.
  • Demonstrated inhibition of Notch-1 activation and tumor cell proliferation.
  • Modulates angiogenesis-related pathways including eNOS, VEGF, and HIF-1α via PI3K-dependent signaling.
  • Storage: keep at 4°C protected from light; in solution, store at -80°C for long-term preservation.
Catalog No. 50-002-89576 Supplier Medchemexpress LLC Supplier No. HYN5112B10MG
β,β-Dimethylacrylshikonin (Isoarnebin I) is a naphthoquinone derivative used in biomedical research for its anticancer and angiogenesis-modulating activities. It is supplied as a solid with high reported purity for laboratory experiments and is typically handled and stored protected from light.
  • High purity (99.3%) appropriate for research use.
  • Solid form with brown to reddish brown appearance.
  • Available in small research pack size (10 mg) for assay development.
  • Demonstrated inhibition of Notch-1 activation and tumor cell proliferation.
  • Modulates angiogenesis-related pathways including eNOS, VEGF, and HIF-1α via PI3K-dependent signaling.
  • Storage: keep at 4°C protected from light; in solution, store at -80°C for long-term preservation.