Learn More
Echelon Biosciences Research Labs DIMETHYLALLYL ThioloDIPHOSPHATE 10 mg
SDP
Supplier:  Echelon Biosciences Research Labs IS05110MG
Encompass
Dimethylallyl thiolodiphosphate (DMASPP) is an analog of DMAPP in which the non-bridging oxygen has been replaced by sulfur resulting in a less-reactive compound. This item is non returnable
Catalog No. 50-720-411