Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylamine, 26% (w/v), Ricca Chemical
SDP

Catalog No. R25691344C1 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Safety-coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R25691344C1 4 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R25691344C1 Supplier Ricca Chemical Company Supplier No. R25691344C1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 124-40-3
Molecular Formula C2H7N
Molecular Weight (g/mol) Mixture
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
Synonym dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
PubChem CID 674
ChEBI CHEBI:17170
IUPAC Name N-methylmethanamine
SMILES CNC

Specifications

CAS Min % 71.11, 28.89
CAS Max % 71.11, 28.89
Color Colorless to Yellow
Density 0.9
pH Alkaline
Flash Point -18°C
Odor Ammonia-Like
Packaging Safety-coated Glass Bottle
Quantity 4 L
Solubility Information Miscible in water
Percent Purity 26% w/v
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Dimethylamine, 26% (w/v)
Show More Show Less
ShelfLife 18 months
Recommended Storage Ambient Flammble
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.