Learn More
Medchemexpress LLC Dimethylamine-d6 hydrochloride | 53170-19-7 | MFCD00064544 | 98.0% | 87.58 | 250mg
SDP
Catalog No. 5000267783
Encompass_Preferred
Dimethylamine-d6 hydrochloride is a deuterium labeled Dimethylamine hydrochloride Dimethylamine hydrochloride is the immediate precursor of dimethylnitrosamine a known potent carcinogen in a wide variety of animal species[1]
Catalog No. 50-002-67783 Supplier Medchemexpress LLC Supplier No. HYY1354S250MG
Dimethylamine-d6 hydrochloride is a deuterium labeled Dimethylamine hydrochloride Dimethylamine hydrochloride is the immediate precursor of dimethylnitrosamine a known potent carcinogen in a wide variety of animal species[1]