Learn More
Medchemexpress LLC Dimethylamine-d6 hydrochloride | 53170-19-7 | MFCD00064544 | 98.0% | 87.58 | C2H2D6ClN | 100mg
SDP
Catalog No. 5000266665
Encompass_Preferred
Dimethylamine-d6 hydrochloride is a deuterium labeled Dimethylamine hydrochloride Dimethylamine hydrochloride is the immediate precursor of dimethylnitrosamine a known potent carcinogen in a wide variety of animal species[1]
Catalog No. 50-002-66665 Supplier Medchemexpress LLC Supplier No. HYY1354S100MG
Dimethylamine-d6 hydrochloride is a deuterium labeled Dimethylamine hydrochloride Dimethylamine hydrochloride is the immediate precursor of dimethylnitrosamine a known potent carcinogen in a wide variety of animal species[1]