Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethylamine Solution

Catalog No. 426458100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-645-8100ML 100 mL
42-645-82L 2 L
42-645-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 42-645-8100ML Supplier Sigma Aldrich Supplier No. 426458100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethylamine (DMA) solution can be used: To synthesize Ge7O14.5F2,[(CH3)2NH2]3(H2O)0.86, a germanate-based porous inclusion complex.To synthesize ethyl 2-dimethylamino-4,6-diarylpyrimidine-5-carboxylates.To convert 5'sa-bromo-2,2'sa-bithiophene-5carboxaldehyde to 5'sa-dimethylamino-2,2'sa-bithiophene-5-carboxaldehyde.

Specifications

CAS 124-40-3
Density 0.89 g/mL at 25°C
Molecular Formula C2H7N
Linear Formula (CH3)2NH
Refractive Index n20/D 1.37
Quantity 100 mL
MDL Number MFCD00008288
Synonym DMA
Molecular Weight (g/mol) 45.08 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.