Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethylamine Solution

Catalog No. 391956800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-195-6800ML 800 mL
39-195-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 39-195-6800ML Supplier Sigma Aldrich Supplier No. 391956800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethylamine solution (2M in THF) has been used in the synthesis of 2-dimethylamino-1-(3,4-methylenedioxyphenyl)propan-1-one (bk-MDDMA) , a +a-keto derivative of 3,4-methylenedioxyamphetamine (MDA) by reacting with 2-bromo-3'sa,4'sa-methylenedioxypropiophenone. It may also be used to synthesize organic intermediates such as dimethyl-(4-nitro-benzyl)-amine, dimethyl-(4-methyl-benzyl)-amine and 4-dimethylamino-but-2-enoic acid [4-(3,4-dichloro-6-fluoro-phenylamino)-quinazolin-6-yl]-amide.

Specifications

CAS 124-40-3
Density 0.85 g/mL at 25°C
Boiling Point ∼60°C
Molecular Formula C2H7N
Linear Formula (CH3)2NH
MDL Number MFCD00008288
Quantity 800 mL
Molecular Weight (g/mol) 45.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.