Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethylamine Solution

Catalog No. 388246100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-824-6100ML 100 mL
38-824-625ML 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 38-824-6100ML Supplier Sigma Aldrich Supplier No. 388246100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethylamine, a secondary amine, is used as a raw material for the production of many agrichemicals and pharmaceuticals. The solvents dimethylformamide (DMF) and dimethylacetamide (DMA) are derived from dimethylamine. It is also used as a precursor for the preparation of amido derivatives.Dimethylamine can be used as a precursor for the preparation of a number of chemical building blocks such as dimethyl(3-chloropropyl)amine , 2-chloro-N,N-dimethylethanimidamide , and 4, 6-dichloro-2-(dimethylamino)-1,3,5-triazine .Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 124-40-3
Density 0.775 g/mL at 25°C
Molecular Formula C2H7N
Linear Formula (CH3)2NH
MDL Number MFCD00008288
Quantity 100 mL
Molecular Weight (g/mol) 45.08 g/mol
Biological Activity Eyes, Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.