Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylaminoacetyl chloride hydrochloride, tech. 85%

Catalog No. AAL1847506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1847506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1847506 Supplier Thermo Scientific Chemicals Supplier No. L1847506
  • $144.00
    $122.65 / Each of 1

    Save $21.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 60853-81-8 | C4H9Cl2NO | 158.02 g/mol

Dimethylaminoacetyl chloride hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylaminoacetyl chloride hydrochloride is used as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store un
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60853-81-8
Molecular Formula C4H9Cl2NO
Molecular Weight (g/mol) 158.02
MDL Number MFCD02094025
InChI Key MVAXDKDOPWPFML-UHFFFAOYSA-N
Synonym 2-dimethylamino acetyl chloride hydrochloride, dimethylaminoacetyl chloride hydrochloride, n,n-dimethylglycinyl chloride hydrochloride, acmc-1b50m, dimethylaminoacetyl chloride-hcl, dimethylamino-acetyl chloride hcl, dimethylaminoacetylchloride hydrochloride, n,n-dimethylglycyl chloride hydrochloride, dimethylamino-acetyl chloride hydrochloride, 2-dimethylaminoacetyl chloride hydrochloride
PubChem CID 13994991
IUPAC Name 2-(dimethylamino)acetyl chloride;hydrochloride
SMILES Cl.CN(C)CC(Cl)=O

Specifications

Quantity 5 g
UN Number UN3261
Beilstein 3980151
Sensitivity Moisture sensitive
Formula Weight 158.03
Percent Purity 85%
Grade Technical
Chemical Name or Material Dimethylaminoacetyl chloride hydrochloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.