Learn More
Cayman Chemical Dimethylamino-Micheliolide, Purity: ≥95%, CAS: 1403357-81-2, 10 mg
SDP
Catalog No. 503015950
Encompass_Preferred
A prodrug form of micheliolide; inhibits migration and invasion of, and vascular formation by, U-118 MG glioma cells at 10 µM; enhances cisplatin-induced cell death in U-118 MG cells; reduces tumor volume and slows the tumor growth rate in a U-118 MG mouse xenograft model at 200 mg/kg when administered in combination with cisplatin; prevents proteinuria and renal failure in a db/db mouse model of diabetic nephropathy
Catalog No. 50-301-5950 Supplier Cayman Chemical Supplier No. 2549910 MG
A prodrug form of micheliolide; inhibits migration and invasion of, and vascular formation by, U-118 MG glioma cells at 10 µM; enhances cisplatin-induced cell death in U-118 MG cells; reduces tumor volume and slows the tumor growth rate in a U-118 MG mouse xenograft model at 200 mg/kg when administered in combination with cisplatin; prevents proteinuria and renal failure in a db/db mouse model of diabetic nephropathy