Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethylcarbamyl chloride

Catalog No. 501860334
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-186-0334 100 g
50-186-0335 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-186-0334 Supplier Sigma Aldrich Supplier No. D152803100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethylcarbamyl chloride
TRUSTED_SUSTAINABILITY

Specifications

CAS 79-44-7
Melting Point -33°C (lit.)
Density 1.168 g/mL (at 25°C (literature))
Boiling Point 167°C to 168°C (775 mmHg)
Molecular Formula C3H6ClNO
Refractive Index n20/D 1.453 (literature)
Linear Formula (CH3)2 NCOCl
MDL Number MFCD00000635
Quantity 100 g
Synonym Chloroformic acid dimethyl amide; Dimethylcarbamoyl chloride
Molecular Weight (g/mol) 107.54
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.