Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyldioctadecylammoniu50G
Catalog No. 502903251
Encompass
Dimethyldioctadecylammonium bromide is a cationic lipid that stimulates immune responses activates macrophages and binds to antigens.
Catalog No. 50-290-3251 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D277950G
Dimethyldioctadecylammonium bromide is a cationic lipid that stimulates immune responses activates macrophages and binds to antigens.