Learn More
Medchemexpress LLC Dimethyldioctadecylammonium (bromide) | 3700-67-2 | ≥99.0% | 100 G
SDP
Catalog No. 5000341152
Encompass_Preferred
Dimethyldioctadecylammonium bromide is a synthetic cationic lipid also known as DODAB or DDAB. It features a positively charged ammonium head group and two long hydrophobic tails. These characteristics enable it to form liposomes and lipid-based nanoparticles for efficient genetic material delivery into cells.
  • Used in gene delivery.
  • Used in vaccine development.
  • Applied as a surfactant.
  • Functions as an emulsifier.
  • Utilized in fabric softeners.
Catalog No. 50-003-41152 Supplier Medchemexpress LLC Supplier No. HY131119100G
Dimethyldioctadecylammonium bromide is a synthetic cationic lipid also known as DODAB or DDAB. It features a positively charged ammonium head group and two long hydrophobic tails. These characteristics enable it to form liposomes and lipid-based nanoparticles for efficient genetic material delivery into cells.
  • Used in gene delivery.
  • Used in vaccine development.
  • Applied as a surfactant.
  • Functions as an emulsifier.
  • Utilized in fabric softeners.