Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyldiphenyltin

Catalog No. AA3961706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3961706 5 g
AA3961703 1 g
AA3961714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3961706 Supplier Thermo Scientific Chemicals Supplier No. 03961706
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1080-43-9 | C14H16Sn | 302.992 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1080-43-9
Molecular Formula C14H16Sn
Molecular Weight (g/mol) 302.992
MDL Number MFCD00058812
InChI Key INCQQSKGFIBXAY-UHFFFAOYSA-N
Synonym dimethyldiphenyltin, acmc-1bqk3, stannane, dimethyldiphenyl
PubChem CID 11130460
IUPAC Name dimethyl(diphenyl)stannane
SMILES C[Sn](C)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Quantity 5 g
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 302.97
Physical Form Liquid
Chemical Name or Material Dimethyldiphenyltin
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.