Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyldivinylsilane 98.0+%, TCI America™
SDP

Catalog No. D48665ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48665ML 5 mL
D486625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48665ML Supplier TCI America Supplier No. D48665ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10519-87-6
Molecular Formula C6H12Si
Molecular Weight (g/mol) 112.247
MDL Number MFCD00053855
InChI Key QRHCILLLMDEFSD-UHFFFAOYSA-N
PubChem CID 66339
IUPAC Name bis(ethenyl)-dimethylsilane
SMILES C[Si](C)(C=C)C=C

Specifications

Color Colorless
Boiling Point 82°C
Quantity 5 mL
UN Number 1993
Formula Weight 112.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyldivinylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.