Learn More
Medchemexpress LLC HY-19944 5mg Medchemexpress, Dimethylenastron CAS:863774-58-7 Purity:>98%
SDP
Catalog No. 502082198
Encompass_Preferred
Medchemexpress, HY-19944 5mg Dimethylenastron CAS:863774-58-7 Dimethylenastron is a potent kinesin Eg5 inhibitor, with an IC50 of 200 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2198 Supplier Medchemexpress LLC Supplier No. HY199445MG
Medchemexpress, HY-19944 5mg Dimethylenastron CAS:863774-58-7 Dimethylenastron is a potent kinesin Eg5 inhibitor, with an IC50 of 200 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.