Learn More
Crescent Chemical Co Inc DIMETHYLFORMAMIDE MOL BIO GRDE
SDP
Catalog No. 50248434
Encompass_Preferred
Dimethylformamide molecular biology grade DNase/RNase not detected. Suitable as a solvent for chromogenic substrates used in molecular biology applications. Size - 250ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 39756.01 CAS 68
Catalog No. 50-248-434 Supplier Crescent Chemical Co Inc Supplier No. SE39756.01
Dimethylformamide molecular biology grade DNase/RNase not detected. Suitable as a solvent for chromogenic substrates used in molecular biology applications. Size - 250ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 39756.01 CAS 68