Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylformamide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019436 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9436 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9436 Supplier MilliporeSigma Supelco Supplier No. PHR15533X1.2ML
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Ignition Point 833°F
CAS 68-12-2
Vapor Pressure 2.7 mmHg (20°C)
Vapor Density 2.5 (vs. air)
Density 0.944 g/mL
Flash Point 58°C - Closed Cup
Linear Formula HCON(CH3)2
MDL Number MFCD00003284
Melting Point -61°C
Boiling Point 153°C
Synonym N,N-Dimethylformamide; DMF; NSC 5356
Type Certified Reference Material
Quantity 3 x 1.2 mL
Formula Weight 73.09
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.