Learn More
Medchemexpress LLC HY-N0085 5mg Medchemexpress, Dimethylfraxetin CAS:6035-49-0 Purity:>98%
SDP
Catalog No. 501933384
Encompass_Preferred
Medchemexpress, HY-N0085 5mg Dimethylfraxetin CAS:6035-49-0 Dimethylfraxetin is a Carbonic anhydrase inhibitor, with a K i value of 0.0097 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3384 Supplier Medchemexpress LLC Supplier No. HYN00855MG
Medchemexpress, HY-N0085 5mg Dimethylfraxetin CAS:6035-49-0 Dimethylfraxetin is a Carbonic anhydrase inhibitor, with a K i value of 0.0097 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.