Learn More
Medchemexpress LLC α,α-dimethylglycine-d6 | 50348-93-1 | 99.7% | 109.16 | C4H3D6NO2 | 5mg
SDP
Catalog No. 5000268877
Encompass_Preferred
NSC 16590-d6 is the deuterium labeled NSC 16590 NSC 16590 inhibits the production of endogenous ethylene in the cotyledonary segments of cocklebur
Catalog No. 50-002-68877 Supplier Medchemexpress LLC Supplier No. HYY0124S5MG
NSC 16590-d6 is the deuterium labeled NSC 16590 NSC 16590 inhibits the production of endogenous ethylene in the cotyledonary segments of cocklebur