Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylglyoxime Disodium Salt Octahydrate 97.0+%, TCI America™
SDP

Catalog No. D073025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D073025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D073025G Supplier TCI America Supplier No. D073025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75006-64-3
Molecular Formula C4H22N2Na2O10
Molecular Weight (g/mol) 304.20
MDL Number MFCD00150288
InChI Key IJJDRGUPRATWQX-UHFFFAOYSA-L
Synonym dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate
PubChem CID 71310106
IUPAC Name disodium oxido[3-(oxidoimino)butan-2-ylidene]amine octahydrate
SMILES O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]

Specifications

Color White
Quantity 25 g
Formula Weight 160.08
Percent Purity ≥97.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime Disodium Salt Octahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.