Learn More
Medchemexpress LLC HY-W007894 500mg Medchemexpress, Dimethylmalonic acid CAS:595-46-0 Purity:>98%
Supplier: Medchemexpress LLC HYW007894500MG
Medchemexpress, HY-W007894 500mg Dimethylmalonic acid CAS:595-46-0 Dimethylmalonic acid is a short-chain dicarboxylic acid in human serum. Dimethylmalonic acid is also a volatile organic compound detected in alveolar breath. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.