Learn More
Medchemexpress LLC Dimethylmalonic acid | 595-46-0 | 132.11 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00789410MM/1ML
Encompass_Preferred
Dimethylmalonic acid is a short-chain dicarboxylic acid found in human serum and identified as a volatile organic compound in alveolar breath. It serves as a key human endogenous metabolite.
  • Found in human serum and alveolar breath
  • Acts as a human endogenous metabolite
Catalog No. 50-004-46518