Learn More
Medchemexpress LLC Dimethylmalonic acid | 595-46-0 | 132.11 | 50 G
SDP
Catalog No. 5000446517
Encompass_Preferred
Dimethylmalonic acid is a short-chain dicarboxylic acid found in human serum and is also a volatile organic compound detected in alveolar breath. It is classified as a human endogenous metabolite.
  • Found in human serum
  • Detected in alveolar breath
  • Classified as a human endogenous metabolite
  • Solid, white to off-white appearance
  • Includes ketone, aldehyde, and acid structures
  • Ships at room temperature (continental US)
  • Powder stable for 3 years at -20°C
  • In-solvent stable for 6 months at -80°C
Catalog No. 50-004-46517 Supplier Medchemexpress LLC Supplier No. HYW00789450G
Add to Cart
Add to Cart
Dimethylmalonic acid is a short-chain dicarboxylic acid found in human serum and is also a volatile organic compound detected in alveolar breath. It is classified as a human endogenous metabolite.
  • Found in human serum
  • Detected in alveolar breath
  • Classified as a human endogenous metabolite
  • Solid, white to off-white appearance
  • Includes ketone, aldehyde, and acid structures
  • Ships at room temperature (continental US)
  • Powder stable for 3 years at -20°C
  • In-solvent stable for 6 months at -80°C