Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylmalonic Acid 98.0+%, TCI America™
SDP

Catalog No. D26065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26065G 5 g
D260625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26065G Supplier TCI America Supplier No. D26065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 595-46-0
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00004193
InChI Key OREAFAJWWJHCOT-UHFFFAOYSA-N
Synonym dimethylmalonic acid, 2,2-dimethylmalonic acid, propanedioic acid, dimethyl, dimethylpropanedioic acid, dimethyl-propanedioic acid, 2,2-propanedicarboxylic acid, unii-55981wkx6b, dimethyl-malonic acid, propanedioic acid dimethyl, 2,2-dimethyl-propanedioic acid
PubChem CID 11686
IUPAC Name dimethylpropanedioic acid
SMILES CC(C)(C(O)=O)C(O)=O

Specifications

Melting Point 191°C
Color White
Quantity 5 g
Formula Weight 132.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylmalonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.