Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylnaphthalene (mixture) 80.0+%, TCI America™
SDP

Catalog No. d075325g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D075325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D075325G Supplier TCI America Supplier No. D075325G

Chemical Identifiers

CAS 28804-88-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004120
InChI Key YGYNBBAUIYTWBF-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dimethyl, dimethylnaphthalene, mixture of isomers, 2,6-dmn, 2,6-dimethyl-naphthalene, unii-76u29qw3fm, 2,6-dimethylphthalene, 2,6-dimethylnaphthalen, naphthalene, 2,6-or 2,7-dimethyl, 2.6-dimethylnaphthalene, acmc-1arta
PubChem CID 11387
ChEBI CHEBI:34251
IUPAC Name 2,6-dimethylnaphthalene
SMILES CC1=CC2=C(C=C1)C=C(C=C2)C

Specifications

Color Brown
Quantity 25 g
Formula Weight 156.23
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethylnaphthalene (mixture)
EINECSNumber (4)-0080
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.