Learn More
Chem Service Inc DIMETHYLNITROBENZENE
SDP
Catalog No. NC9514087
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
1, 3-Dimethyl-2-nitrobenzene Solution, Synonyms: 2-Nitro-m-xylene; 2, 6-Dimethylnitrobenzene, CAS No.: 81-20-9, MF: C8H9NO2, MP: -109 deg.C, BP: 55.2 deg.C, SG: 0.7404, FlashPoint: -28 deg.C, Liquid, Slightly Soluble in Water, Hazard: Xn, 5mL
Catalog No. NC9514087 Supplier Chem Service Inc Supplier No. S10114T55ML
This item has a minimum order quantity of 2 per supplier requirements
1, 3-Dimethyl-2-nitrobenzene Solution, Synonyms: 2-Nitro-m-xylene; 2, 6-Dimethylnitrobenzene, CAS No.: 81-20-9, MF: C8H9NO2, MP: -109 deg.C, BP: 55.2 deg.C, SG: 0.7404, FlashPoint: -28 deg.C, Liquid, Slightly Soluble in Water, Hazard: Xn, 5mL