Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride solution

Catalog No. 435694100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-569-4100ML 100 mL
43-569-4500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-569-4100ML Supplier Sigma Aldrich Supplier No. 435694100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride (DTSACl) is an organosilane which can be used as a silica precursor. It forms an oil soluble solution that can act as an emulsion stabilizer.DTSACl may be used for modification of silica.{61} It was used to functionalize Fe3O4/silica-ßcore-shell nanoparticles. {62} DTSACl may be used as a precursor to form silica, 3 and MeMFI zeolite.{63} It acts as a mesopore-generating agent. {64}

Specifications

CAS 27668-52-6
Density 0.88 g/mL (at 25°C)
Molecular Formula C26H58ClNO3Si
Refractive Index n20/D 1.3889
Linear Formula [(CH3O)3 Si(CH2)3 N(CH3)2(CH2)17 CH3]Cl
MDL Number MFCD00054224
Quantity 100 mL
Synonym Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride
Molecular Weight (g/mol) 496.28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.