Learn More
Echelon Biosciences Research Labs DIMETHYLOXALYLGLYCINE 500 mg
SDP
Supplier:  Echelon Biosciences Research Labs F0010500MG
Encompass
Hypoxia Inducible Factor (HIF) regulates responses to hypoxia and is comprised of two subunits alpha and beta. Upon cellular exposure to hypoxic conditions, the HIF complex (alpha andbeta subunits) is stabilized and binds to DNA transcriptionally activating genes linked to the cellular processes of angiogenesis and glucose metabolism1. Under normal conditions, the HIF-alpha subunit is hydroxylated by the enzyme HIF-alpha prolyl hydroxylase (HIF-PH) leading to ubiquitylation of HIF-alpha and subsequent destruction2. DMOG is a cell permeable competitive inhibitor of HIF-alpha prolyl hydroxylase (HIF-PH) leading to the stabilization of HIF and subsequent angiogenesis and glucose metabolism at concentrations between 0.1 and 1 mM3 This item is non returnable
Catalog No. 50-720-322