Learn More
Strem, An Ascensus Company CAS# 672-66-2. 2g. Dimethylphenylphosphine, 99%. MFCD00008509
SDP
Catalog No. 5090110136
Encompass
CAS# 672-66-2. 2g. Dimethylphenylphosphine, 99%. MFCD00008509. Molecular Weight: 138.15. Molecular Formula: (CH3)2(C6H5)P. Color/form: colorless liq. Strem# 15-1400. http://www.strem.com/catalog/v/15-1400/
Catalog No. 50-901-10136 Supplier Strem, An Ascensus Company Supplier No. 1514002G
CAS# 672-66-2. 2g. Dimethylphenylphosphine, 99%. MFCD00008509. Molecular Weight: 138.15. Molecular Formula: (CH3)2(C6H5)P. Color/form: colorless liq. Strem# 15-1400. http://www.strem.com/catalog/v/15-1400/