Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylphenylphosphine, C 69.2%, H 7.9%, Thermo Scientific™

Catalog No. AA3018809
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3018809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3018809 Supplier Thermo Scientific Chemicals Supplier No. 3018809

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-66-2
Molecular Formula C8H11P
Molecular Weight (g/mol) 138.15
MDL Number MFCD00008509
InChI Key HASCQPSFPAKVEK-UHFFFAOYSA-N
Synonym dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine
PubChem CID 69597
ChEBI CHEBI:30671
IUPAC Name dimethyl(phenyl)phosphane
SMILES CP(C)C1=CC=CC=C1

Specifications

Density 0.971g/mL
Boiling Point 75°C to 79°C (12 mmHg)
Odor Unpleasant
Quantity 10 g
UN Number UN1993
Formula Weight 138.15
Concentration or Composition (by Analyte or Components) C 69.2%, H 7.9%
Physical Form Liquid
Chemical Name or Material Dimethylphenylphosphine
Hazard Category Warning
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.