Learn More
Medchemexpress LLC Dimethylpropiothetin 250g
SDP
Supplier:  Medchemexpress LLC 250GHYW105506
Encompass_Preferred
Dimethylpropiothetin hydrochloride (s s-Dimethyl- -propionic acid thetine) is the hydrochloride form of dimethylpropiothetin Dimethylpropiothetin can be isolated in marine algae and is a precursor to dimethylsulfide (DMS) Dimethylpropiothetin is involved in osmoregulation in Hymenomonas carterae[1]
Catalog No. 50-002-06023