Learn More
Strem, An Ascensus Company CAS# 593-79-3. 50g. Dimethylselenide (99.99+%-Se) PURATREM. MFCD00014848
SDP
Catalog No. 5090111447
Encompass
CAS# 593-79-3. 50g. Dimethylselenide (99.99+%-Se) PURATREM. MFCD00014848. Molecular Weight: 109.03. Molecular Formula: (CH3)2Se. Color/form: colorless to yellow liq. Strem# 34-0550. http://www.strem.com/catalog/v/34-0550/
Catalog No. 50-901-11447 Supplier Strem, An Ascensus Company Supplier No. 34055050G
CAS# 593-79-3. 50g. Dimethylselenide (99.99+%-Se) PURATREM. MFCD00014848. Molecular Weight: 109.03. Molecular Formula: (CH3)2Se. Color/form: colorless to yellow liq. Strem# 34-0550. http://www.strem.com/catalog/v/34-0550/