Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylselenide 98.0+%, TCI America™
SDP

Catalog No. D12891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12891G Supplier TCI America Supplier No. D12891G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 593-79-3
Molecular Formula C2H6Se
Molecular Weight (g/mol) 109.041
MDL Number MFCD00014848
InChI Key RVIXKDRPFPUUOO-UHFFFAOYSA-N
Synonym dimethylselenide, dimethylselenium, dimethyl selenide, methyl selenide, methyl selenium, methane, selenobis, selenium dimethyl, selenide, dimethyl, ch3 2se, unii-yk0r6jkt6h
PubChem CID 11648
ChEBI CHEBI:4610
IUPAC Name methylselanylmethane
SMILES C[Se]C

Specifications

Color Yellow
Boiling Point 58°C
Quantity 1 g
UN Number 3440
Formula Weight 109.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethylselenide
RTECSNumber VS6880000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.