Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylsilyldiethylamine, 95%

Catalog No. AAL0350314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0350314 25 g
AAL0350306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0350314 Supplier Thermo Scientific Chemicals Supplier No. L0350314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13686-66-3 | C6H16NSi | 130.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13686-66-3
Molecular Formula C6H16NSi
Molecular Weight (g/mol) 130.29
MDL Number MFCD00048523
InChI Key ADTGAVILDBXARD-UHFFFAOYSA-N
Synonym dimethylsilyldiethylamine, n,n-diethyl-1,1-dimethylsilylamine, diethylamino dimethylsilyl, silanamine, n,n-diethyl-1,1-dimethyl, silanamine,n,n-diethyl-1,1-dimethyl, diethylamino dimethyl silicon, diethylaminodimethylsilane, n,n-diethyl-1,1-dimethylsilanamine
PubChem CID 6335300
IUPAC Name (diethylamino)dimethylsilyl
SMILES CCN(CC)[Si](C)C

Specifications

Boiling Point 109°C to 110°C
Quantity 25 g
UN Number UN1993
Beilstein 741862
Flash Point <−5°C (23°F)
Formula Weight 131.3
Percent Purity 95%
Refractive Index 1.408
Sensitivity Moisture sensitive
Density 0.8
Chemical Name or Material Dimethylsilyldiethylamine
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 237-202-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.