Learn More
Supelco Inc Dimethyl Sulfoxide Biotech, 50 Milliliter
Catalog No. NC9372364
Encompass
Dimethyl sulfoxide; 50mL; M.W. 78.13; (CH3)2SO; B.P. 189 deg.C(lit.); M.P. 16 deg. to 19 deg.C(lit.); CAS: 67-68-5
Catalog No. NC9372364 Supplier Supelco Inc Supplier No. D243850ML
Dimethyl sulfoxide; 50mL; M.W. 78.13; (CH3)2SO; B.P. 189 deg.C(lit.); M.P. 16 deg. to 19 deg.C(lit.); CAS: 67-68-5