Learn More
ATCC Dimethylsulfoxide (DMSO), 5x5ml
Supplier:  ATCC 4X/DEL
Encompass
DMSO is used as a cyroprotectant for the cryopreservation of cell lines. ATCC DMSO is cell culture grade and has been tested to assure nontoxicity and sterility. Store DMSO at 2°C to 8°C when not in use. It may be solid at this temperature. However, it may also be stored at room temperature where it remains a liquid. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50188278NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-278FP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product