Learn More
ATCC Dimethylsulfoxide (DMSO), 5 x 5.0 mL, Sterile, Cell Culture Tested
Catalog No. 502383363
Encompass
Dimethylsulfoxide (DMSO) is used as a cyroprotectant for the cryopreservation of cell lines. ATCC DMSO is cell culture grade and has been tested to assure nontoxicity and sterility. DMSO has been filtered through a 0.2 ?m PTFE membrane and aseptically dispensed. Each amber vial contains a 5 mL sterile aliquot. This product has applications for cyropreservation, cell culture, cell growth, and viability.
Catalog No. 50-238-3363 Supplier ATCC Supplier No. 4X
 Restricted Item
Add to Cart
Add to Cart
Dimethylsulfoxide (DMSO) is used as a cyroprotectant for the cryopreservation of cell lines. ATCC DMSO is cell culture grade and has been tested to assure nontoxicity and sterility. DMSO has been filtered through a 0.2 ?m PTFE membrane and aseptically dispensed. Each amber vial contains a 5 mL sterile aliquot. This product has applications for cyropreservation, cell culture, cell growth, and viability.