Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylthiophosphinoyl Chloride 97.0+%, TCI America™
SDP

Catalog No. D21595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21595G 5 g
D21591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21595G Supplier TCI America Supplier No. D21595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 993-12-4
Molecular Formula C2H6ClPS
Molecular Weight (g/mol) 128.55
MDL Number MFCD00014454
InChI Key UGOLAPHJCTVIEW-UHFFFAOYSA-N
Synonym Chlorodimethylphosphine Sulfide, Dimethylphosphinothioyl Chloride
PubChem CID 61245
IUPAC Name chlorodimethyl-λ⁵-phosphanethione
SMILES CP(C)(Cl)=S

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
UN Number 3265
Formula Weight 128.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethylthiophosphinoyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.